siRNA Details
Gene Symbol: Fam173a | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001285982.1 | NP_001272911.1 | 5 | 476 | ||
NM_001285984.1 | NP_001272913.1 | 5 | 476 | |||
NM_145410.4 | NP_663385.2 | 5 | 761 | |||
GenBank mRNA | AK146214.1 | - | 5 | 742 | ||
AK162337.1 | - | 5 | 737 | |||
BC008155.1 | - | 5 | 485 | 3154632 | ||
BC096050.1 | - | 5 | 656 | 5119802 |
