siRNA Details
| Gene Symbol: Fbxl16 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001164225.1 | NP_001157697.1 | 6 | 2477 | ||
| GenBank mRNA | BC042620.1 | - | 5 | 2186 | ||
| BC132383.1 | - | 5 | 2273 | 40130660 | ||
| BC137657.1 | - | 5 | 2273 | 8860676 | ||

Fbxl16
Genomic Map