siRNA Details
| Gene Symbol: Mob3b | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_178061.5 | NP_835162.1 | 2 | 755 | ||
| GenBank mRNA | AK039841.1 | - | 2 | 601 | ||
| AK087285.1 | - | 2 | 756 | |||
| AK155217.1 | - | 2 | 756 | |||
| AK170973.1 | - | 2 | 749 | |||
| BC020028.1 | - | 2 | 737 | 4007373 | ||

Mob3b
Genomic Map