siRNA Details
| Gene Symbol: Rhot2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_145999.2 | NP_666111.1 | 17 | 1913 | ||
| GenBank mRNA | AK142183.1 | - | 12 | 2019 | ||
| AK216218.1 | - | 3 | 257 | |||
| AY311390.1 | - | 17 | 1913 | |||
| BC023163.1 | - | 7 | 683 | |||
| BC023927.1 | - | 3 | 966 | |||
| BC029777.1 | - | 17 | 1546 | 4950061 | ||
| BC034062.1 | - | 17 | 1568 | 4207654 | ||

Rhot2
Genomic Map