siRNA Details
| Gene Symbol: Vezt | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| GenBank mRNA | AK031848.1 | - | 12 | 2007 | ||
| AK036017.1 | - | 12 | 1944 | |||
| AY753561.1 | - | 12 | 1900 | |||
| BC056428.1 | - | 12 | 1931 | 5716245 | ||
Please enter the information below and press OK to send your cart to Core Services for purchase.

Vezt| Gene Symbol: Vezt | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| GenBank mRNA | AK031848.1 | - | 12 | 2007 | ||
| AK036017.1 | - | 12 | 1944 | |||
| AY753561.1 | - | 12 | 1900 | |||
| BC056428.1 | - | 12 | 1931 | 5716245 | ||