siRNA Details
| Gene Symbol: Fam20b | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_145413.4 | NP_663388.1 | 8 | 1192 | ||
| GenBank mRNA | AK049705.1 | - | 7 | 994 | ||
| AK129154.1 | - | 6 | 601 | |||
| AK132149.1 | - | 8 | 1193 | |||
| BC019381.1 | - | 8 | 1211 | 4206622 | ||
| BC023737.1 | - | 9 | 1408 | 5352660 | ||
| BC024412.1 | - | 7 | 1142 | 4971483 | ||
| BC031473.1 | - | 8 | 1304 | 3990487 | ||

Fam20b
Genomic Map