siRNA Details
| Gene Symbol: Bud13 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_146000.2 | NP_666112.1 | 6 | 1347 | ||
| GenBank mRNA | AK037134.1 | - | 6 | 1344 | ||
| AK038126.1 | - | 6 | 1352 | |||
| AK051047.1 | - | 6 | 1352 | |||
| AK077744.1 | - | 6 | 1351 | |||
| BC025490.1 | - | 6 | 1348 | 5252666 | ||

Bud13
Genomic Map