siRNA Details
| Gene Symbol: Slc43a2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001199283.1 | NP_001186212.1 | 8 | 1101 | ||
| NM_001199284.1 | NP_001186213.1 | 8 | 883 | |||
| NM_173388.2 | NP_775564.1 | 8 | 846 | |||
| GenBank mRNA | AB120363.1 | - | 8 | 879 | ||
| AK143773.1 | - | 8 | 944 | |||
| AK159144.1 | - | 8 | 1069 | |||
| AK171104.1 | - | 8 | 1102 | |||
| AK172685.1 | - | 8 | 873 | |||
| BC042513.1 | - | 8 | 842 | 3964020 | ||

Slc43a2
Genomic Map