siRNA Details
| Gene Symbol: Slc36a3 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_172258.3 | NP_758462.2 | 8 | 1056 | ||
| GenBank mRNA | AF453745.1 | - | 8 | 1056 | ||
| AK132910.1 | - | 9 | 1142 | |||
| AY211261.1 | - | 8 | 1038 | |||
Please enter the information below and press OK to send your cart to Core Services for purchase.

Slc36a3| Gene Symbol: Slc36a3 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_172258.3 | NP_758462.2 | 8 | 1056 | ||
| GenBank mRNA | AF453745.1 | - | 8 | 1056 | ||
| AK132910.1 | - | 9 | 1142 | |||
| AY211261.1 | - | 8 | 1038 | |||