siRNA Details
| Gene Symbol: Ncaph | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_144818.3 | NP_659067.2 | 6 | 760 | ||
| GenBank mRNA | AK028919.1 | - | 6 | 745 | ||
| AK042706.1 | - | 6 | 677 | |||
| AK080433.1 | - | 6 | 749 | |||
| AK129047.1 | - | 3 | 313 | |||
| BC021499.1 | - | 6 | 734 | 5346298 | ||
| BC025060.1 | - | 6 | 685 | 3673890 | ||

Ncaph
Genomic Map