siRNA Details
| Gene Symbol: Rnpep | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001159624.1 | NP_001153096.1 | 6 | 1209 | ||
| NM_145417.3 | NP_663392.2 | 7 | 1326 | |||
| GenBank mRNA | AK031138.1 | - | 6 | 1149 | ||
| AK145466.1 | - | 7 | 1329 | |||
| AK159195.1 | - | 7 | 1269 | |||
| AK159445.1 | - | 7 | 1258 | |||
| AK159625.1 | - | 7 | 1269 | |||
| BC010520.1 | - | 7 | 1287 | |||
| BC019200.1 | - | 7 | 1297 | 5041005 | ||

Rnpep
Genomic Map