siRNA Details
| Gene Symbol: Nav1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_173437.2 | NP_775613.2 | 28, 29 | 5370 | ||
| GenBank mRNA | AF307453.1 | - | 28, 29 | 5730 | ||
| AK036761.1 | - | 10, 11 | 1374 | |||
| AK122458.1 | - | 28, 29 | 5428 | |||
| AK153742.1 | - | 11, 12 | 1691 | |||
| BC080292.1 | - | 10, 11 | 1442 | |||

Nav1
Genomic Map