siRNA Details
Gene Symbol: Adgb | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001127353.2 | NP_001120825.2 | 13 | 1561 | ||
GenBank mRNA | AK078911.1 | - | 4 | 542 | ||
AK134038.1 | - | 2 | 102 |

Please enter the information below and press OK to send your cart to Core Services for purchase.
Gene Symbol: Adgb | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001127353.2 | NP_001120825.2 | 13 | 1561 | ||
GenBank mRNA | AK078911.1 | - | 4 | 542 | ||
AK134038.1 | - | 2 | 102 |