siRNA Details
| Gene Symbol: Adgb | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001127353.2 | NP_001120825.2 | 18 | 2044 | ||
| GenBank mRNA | AK078911.1 | - | 9 | 1019 | ||
Please enter the information below and press OK to send your cart to Core Services for purchase.

Adgb| Gene Symbol: Adgb | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001127353.2 | NP_001120825.2 | 18 | 2044 | ||
| GenBank mRNA | AK078911.1 | - | 9 | 1019 | ||