siRNA Details
| Gene Symbol: PFDN4 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_002623.3 | NP_002614.2 | 3 | 327 | ||
| XM_017027879.1 | XP_016883368.1 | 3 | 611 | |||
| GenBank mRNA | AK312059.1 | - | 3 | 203 | ||
| BC010953.1 | - | 3 | 207 | 4273238 | ||
| CN482830.1 | - | 3 | 327 | |||
| U41816.1 | - | 3 | 188 | |||

PFDN4
Genomic Map