siRNA Details
| Gene Symbol: Gm4794 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001101452.1 | NP_001094922.1 | 2 | 315 | ||
| Gene Symbol: Sult3a1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_020565.2 | NP_065590.2 | 3 | 376 | ||
| GenBank mRNA | AF026075.1 | - | 3 | 377 | ||
| BC125593.1 | - | 3 | 376 | 40130108 | ||
| BC125595.1 | - | 3 | 376 | 40130110 | ||

Gm4794
Genomic Map