siRNA Details
| Gene Symbol: Lace1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_145743.2 | NP_665686.2 | 4 | 623 | ||
| GenBank mRNA | AF397909.1 | - | 3 | 326 | ||
| AF520417.1 | - | 4 | 472 | |||
| AK044595.1 | - | 4 | 499 | |||
| AK170996.1 | - | 4 | 474 | |||
| BC023308.1 | - | 3 | 300 | |||
| BC089595.1 | - | 4 | 473 | 30118411 | ||

Lace1
Genomic Map