siRNA Details
| Gene Symbol: Mcu | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001033259.4 | NP_001028431.2 | 8 | 1014 | ||
| GenBank mRNA | AK161322.1 | - | 5 | 836 | ||
| BC139254.1 | - | 5 | 631 | 8862276 | ||
| BC139257.1 | - | 5 | 631 | 8862279 | ||
| DV649183.1 | - | 3 | 285 | |||

Mcu
Genomic Map