siRNA Details
| Gene Symbol: Zfp365 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_178679.2 | NP_848794.1 | 2 | 800 | ||
| GenBank mRNA | AF543761.1 | - | 1 | 521 | ||
| AJ516033.1 | - | 2 | 662 | |||
| AK031846.1 | - | 2 | 596 | |||
| AK033892.1 | - | 2 | 562 | |||
| AK044322.1 | - | 2 | 559 | |||
| AK077316.1 | - | 2 | 573 | |||
| AK122387.1 | - | 2 | 787 | |||
| BC080272.1 | - | 2 | 760 | 6837597 | ||
| BC157983.1 | - | 2 | 601 | 9088140 | ||
| BC157991.1 | - | 2 | 601 | 9088144 | ||

Zfp365
Genomic Map