siRNA Details
| Gene Symbol: Ilvbl | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_173751.4 | NP_776112.1 | 14, 15 | 1783 | ||
| GenBank mRNA | AK087956.1 | - | 13, 14 | 1657 | ||
| AK146075.1 | - | 13, 14 | 1711 | |||
| AK169899.1 | - | 13, 14 | 1709 | |||
| BC056459.1 | - | 14, 15 | 1741 | 5718157 | ||

Ilvbl
Genomic Map