siRNA Details
| Gene Symbol: Vmn2r81 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_175936.1 | NP_787950.1 | 1, 2 | 248 | ||
| GenBank mRNA | AJ543404.1 | - | 1, 2 | 248 | ||
More Information
Set Membership: |
|
Please enter the information below and press OK to send your cart to Core Services for purchase.

Vmn2r81| Gene Symbol: Vmn2r81 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_175936.1 | NP_787950.1 | 1, 2 | 248 | ||
| GenBank mRNA | AJ543404.1 | - | 1, 2 | 248 | ||
Set Membership: |
|