siRNA Details
Gene Symbol: Vmn2r81 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_175936.1 | NP_787950.1 | 6 | 2223 | ||
GenBank mRNA | AJ543404.1 | - | 6 | 2223 |

More Information
Set Membership: |
![]() |
Panther Classification:
Gene Ontology Categories:
