siRNA Details
| Gene Symbol: Med16 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001163276.1 | NP_001156748.1 | 11 | 1928 | ||
| NM_198107.2 | NP_932775.2 | 11 | 1931 | |||
| GenBank mRNA | AK042340.1 | - | 11 | 1893 | ||
| AK149345.1 | - | 11 | 1912 | |||
| BC057056.1 | - | 11 | 1885 | 6813010 | ||

Med16
Genomic Map