siRNA Details
| Gene Symbol: Sbno2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_183426.1 | NP_906271.1 | 26 | 3122 | ||
| GenBank mRNA | AJ512611.1 | - | 27 | 3278 | ||
| AK149806.1 | - | 26 | 3146 | |||
| AK155462.1 | - | 26 | 3120 | |||
| AK155674.1 | - | 26 | 3119 | |||
| BC032886.1 | - | 6 | 796 | |||
| BC056369.1 | - | 26 | 3122 | 6831600 | ||

Sbno2
Genomic Map