siRNA Details
| Gene Symbol: Sbno2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_183426.1 | NP_906271.1 | 18 | 2118 | ||
| GenBank mRNA | AJ512611.1 | - | 19 | 2274 | ||
| AK089496.1 | - | 14 | 2038 | |||
| AK149806.1 | - | 18 | 2142 | |||
| AK155462.1 | - | 18 | 2116 | |||
| AK155674.1 | - | 18 | 2115 | |||
| BC051234.1 | - | 1 | 211 | |||
| BC056369.1 | - | 18 | 2118 | 6831600 | ||
| BC064113.1 | - | 15 | 1875 | |||

Sbno2
Genomic Map