siRNA Details
| Gene Symbol: Dos | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001195268.1 | NP_001182197.1 | 9 | 1898 | ||
| GenBank mRNA | AK137009.1 | - | 11 | 2235 | ||
| BC024795.1 | - | 1 | 1125 | |||
| BC026893.1 | - | 1 | 1339 | |||
| BC039779.1 | - | 1 | 1062 | |||
| BC052075.1 | - | 10 | 1952 | |||
| BC078442.1 | - | 5 | 1453 | |||
| BC100552.1 | - | 4 | 1281 | 5687195 | ||

Dos
Genomic Map