siRNA Details
| Gene Symbol: AU041133 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001163064.1 | NP_001156536.1 | 4 | 941 | ||
| GenBank mRNA | AK137251.1 | - | 4 | 941 | ||
Please enter the information below and press OK to send your cart to Core Services for purchase.

AU041133| Gene Symbol: AU041133 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001163064.1 | NP_001156536.1 | 4 | 941 | ||
| GenBank mRNA | AK137251.1 | - | 4 | 941 | ||