siRNA Details
Gene Symbol: AU041133 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001163064.1 | NP_001156536.1 | 4 | 423 | ||
GenBank mRNA | AK137251.1 | - | 4 | 423 |

Please enter the information below and press OK to send your cart to Core Services for purchase.
Gene Symbol: AU041133 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001163064.1 | NP_001156536.1 | 4 | 423 | ||
GenBank mRNA | AK137251.1 | - | 4 | 423 |