siRNA Details
| Gene Symbol: Aldh1l2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_153543.2 | NP_705771.2 | 22 | 2738 | ||
| GenBank mRNA | AK052361.1 | - | 22 | 2629 | ||
| AK141954.1 | - | 22 | 2699 | |||
| AK145046.1 | - | 3 | 285 | |||
| BC034531.1 | - | 22 | 2660 | 4188055 | ||

Aldh1l2
Genomic Map