siRNA Details
| Gene Symbol: Ckap4 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_175451.1 | NP_780660.1 | 2 | 1640 | ||
| GenBank mRNA | AK030708.1 | - | 2 | 1524 | ||
| AK089935.1 | - | 1 | 886 | |||
| AK188225.1 | - | 1 | 200 | |||
| BC025522.1 | - | 2 | 1291 | |||
| BC138318.1 | - | 3 | 1662 | 8861338 | ||
| BC138320.1 | - | 3 | 1662 | 8861340 | ||

Ckap4
Genomic Map