siRNA Details
| Gene Symbol: Tcp11l2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_146008.2 | NP_666120.1 | 8 | 1306 | ||
| GenBank mRNA | AK088802.1 | - | 4 | 629 | ||
| AK146035.1 | - | 8 | 1631 | |||
| BC008239.1 | - | 7 | 1102 | |||
| BC029663.1 | - | 8 | 1306 | 5364434 | ||

Tcp11l2
Genomic Map