siRNA Details
| Gene Symbol: Eea1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001001932.3 | NP_001001932.1 | 13 | 1715 | ||
| GenBank mRNA | AK046231.1 | - | 13 | 1624 | ||
| AK082278.1 | - | 12 | 1560 | |||
| AK163235.1 | - | 13 | 1650 | |||
| BC021363.1 | - | 14 | 1802 | |||
| BC075637.1 | - | 13 | 1628 | 30545051 | ||

Eea1
Genomic Map