siRNA Details
| Gene Symbol: March9 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001033262.2 | NP_001028434.1 | 4 | 1764 | ||
| GenBank mRNA | AK158026.1 | - | 4 | 1767 | ||
| BC019560.1 | - | 3 | 931 | |||
Please enter the information below and press OK to send your cart to Core Services for purchase.

March9| Gene Symbol: March9 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001033262.2 | NP_001028434.1 | 4 | 1764 | ||
| GenBank mRNA | AK158026.1 | - | 4 | 1767 | ||
| BC019560.1 | - | 3 | 931 | |||