siRNA Details
| Gene Symbol: Ccm2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001190343.1 | NP_001177272.1 | 4 | 482 | ||
| NM_001190344.1 | NP_001177273.1 | 5 | 568 | |||
| NM_146014.3 | NP_666126.1 | 5 | 656 | |||
| GenBank mRNA | AK155145.1 | - | 5 | 630 | ||
| AK166131.1 | - | 2 | 183 | |||
| AK169727.1 | - | 5 | 570 | |||
| AY442689.1 | - | 5 | 543 | |||
| BC029157.1 | - | 5 | 587 | 4952288 | ||

Ccm2
Genomic Map