siRNA Details
| Gene Symbol: Aftph | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001252503.2 | NP_001239432.1 | 2 | 1626 | ||
| NM_001290545.1 | NP_001277474.1 | 2 | 1626 | |||
| NM_181411.4 | NP_852076.1 | 2 | 1626 | |||
| GenBank mRNA | AK029681.1 | - | 2 | 1525 | ||
| AK030572.1 | - | 2 | 1570 | |||
| AK047225.1 | - | 1 | 690 | |||
| AK136815.1 | - | 1 | 878 | |||
| AK138793.1 | - | 2 | 1548 | |||
| BC052036.1 | - | 2 | 1554 | 5702534 | ||

Aftph
Genomic Map