siRNA Details
| Gene Symbol: Wdpcp | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_145425.3 | NP_663400.2 | 10 | 1027 | ||
| GenBank mRNA | AF424701.1 | - | 10 | 990 | ||
| AK043672.1 | - | 9 | 914 | |||
| AK083042.1 | - | 10 | 1028 | |||
| BC141166.1 | - | 10 | 991 | 9055778 | ||

Wdpcp
Genomic Map