siRNA Details
| Gene Symbol: 4931440F15Rik | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_176829.2 | NP_789799.1 | 1 | 1553 | ||
| GenBank mRNA | AK029918.1 | - | 1 | 1553 | ||
| BC139107.1 | - | 1 | 1410 | 8862129 | ||
| BC139123.1 | - | 1 | 1410 | 8862145 | ||

4931440F15Rik
Genomic Map