siRNA Details
| Gene Symbol: Adamts2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001277305.1 | NP_001264234.1 | 4 | 876 | ||
| NM_175643.3 | NP_783574.1 | 4 | 876 | |||
| GenBank mRNA | AK220464.1 | - | 3 | 618 | ||
| BC046456.1 | - | 4 | 876 | 3965027 | ||

Adamts2
Genomic Map