siRNA Details
| Gene Symbol: Mfap3 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_145426.2 | NP_663401.2 | 3 | 583 | ||
| NM_180599.1 | NP_850930.1 | 3 | 547 | |||
| GenBank mRNA | AK042988.1 | - | 3 | 531 | ||
| AK076071.1 | - | 3 | 549 | |||
| AK155122.1 | - | 3 | 561 | |||
| BC006828.1 | - | 3 | 542 | 3597878 | ||
| BC058567.1 | - | 1 | 1670 | |||
| CF174749.1 | - | 3 | 547 | |||

Mfap3
Genomic Map