siRNA Details
| Gene Symbol: Mrpl22 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_175001.3 | NP_778166.2 | 4 | 245 | ||
| GenBank mRNA | AK077615.1 | - | 4 | 239 | ||
| AK086913.1 | - | 4 | 224 | |||
| AK164417.1 | - | 4 | 224 | |||
| BC061191.1 | - | 4 | 235 | 6703228 | ||
| BY790081.1 | - | 4 | 246 | |||

Mrpl22
Genomic Map