siRNA Details
| Gene Symbol: Wnt9a | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_139298.2 | NP_647459.1 | 3 | 647 | ||
| GenBank mRNA | AB072311.1 | - | 3 | 611 | ||
More Information
Other Species Targeted
This siRNA also targets the following transcripts in Rat: NM_001105783, XM_006246448.

Wnt9a
Genomic Map