siRNA Details
| Gene Symbol: Tom1l2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001039092.3 | NP_001034181.1 | 5 | 521 | ||
| NM_001039093.1 | NP_001034182.1 | 5 | 521 | |||
| NM_153080.2 | NP_694720.2 | 5 | 521 | |||
| GenBank mRNA | AF467887.1 | - | 5 | 501 | ||
| AK036788.1 | - | 5 | 497 | |||
| AK043095.1 | - | 5 | 501 | |||
| AK054531.1 | - | 5 | 489 | |||
| AK166040.1 | - | 5 | 521 | |||
| AK206463.1 | - | 2 | 175 | |||
| AK206592.1 | - | 2 | 175 | |||
| AK218883.1 | - | 2 | 175 | |||
| BC062947.1 | - | 5 | 485 | 30536491 | ||
| BC127266.1 | - | 5 | 487 | 30357295 | ||

Tom1l2
Genomic Map