siRNA Details
| Gene Symbol: Kdm6b | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001017426.1 | NP_001017426.1 | 19 | 4749 | ||
| GenBank mRNA | AK129121.1 | - | 9 | 1541 | ||
| AK136019.1 | - | 9 | 930 | |||
| BC038313.1 | - | 4 | 387 | |||
| BC075632.1 | - | 20 | 4925 | 30362899 | ||

Kdm6b
Genomic Map