siRNA Details
| Gene Symbol: Taok1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_144825.2 | NP_659074.2 | 14 | 2180 | ||
| GenBank mRNA | AK143530.1 | - | 14 | 2187 | ||
| BC016522.1 | - | 2 | 174 | |||
| BC047271.1 | - | 13 | 1527 | |||
| BC151012.1 | - | 14 | 1974 | 9087925 | ||
| BC151015.1 | - | 14 | 1974 | 9087928 | ||
| BC151016.1 | - | 14 | 1973 | 9087929 | ||
| CN718413.1 | - | 4 | 515 | |||
| CN724388.1 | - | 2 | 273 | |||

Taok1
Genomic Map