siRNA Details
| Gene Symbol: Evi2b | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001077496.1 | NP_001070964.1 | 2 | 1082 | ||
| GenBank mRNA | AK039968.1 | - | 2 | 1059 | ||
| AK170476.1 | - | 2 | 1044 | |||
| AK171686.1 | - | 2 | 999 | |||
| BC017548.1 | - | 2 | 1031 | 4922351 | ||
| Gene Symbol: Gm21975 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_146023.4 | NP_666135.1 | 4 | 1517 | ||

Evi2b
Genomic Map