siRNA Details
| Gene Symbol: Heatr6 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_145432.3 | NP_663407.2 | 3 | 465 | ||
| GenBank mRNA | AK012216.1 | - | 3 | 409 | ||
| AK076219.1 | - | 3 | 593 | |||
| AK132494.1 | - | 3 | 400 | |||
| AK149059.1 | - | 3 | 259 | |||
| AK158323.1 | - | 3 | 595 | |||
| AK169901.1 | - | 3 | 412 | |||
| BC065092.1 | - | 3 | 391 | 6855410 | ||
| DV657360.1 | - | 3 | 465 | |||

Heatr6
Genomic Map