siRNA Details
| Gene Symbol: Heatr6 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_145432.3 | NP_663407.2 | 20 | 3406 | ||
| GenBank mRNA | AK076219.1 | - | 20 | 3535 | ||
| AK132494.1 | - | 20 | 3345 | |||
| AK136845.1 | - | 4 | 757 | |||
| AK138219.1 | - | 4 | 664 | |||
| BC003942.1 | - | 13 | 2121 | 3487155 | ||
| BC065092.1 | - | 20 | 3332 | 6855410 | ||

Heatr6
Genomic Map