siRNA Details
| Gene Symbol: Tada2a | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_172562.3 | NP_766150.1 | 13 | 1025 | ||
| GenBank mRNA | AK083467.1 | - | 2 | 56 | ||
| AK157938.1 | - | 13 | 1018 | |||
| AK177030.1 | - | 3 | 187 | |||
| AK209681.1 | - | 3 | 187 | |||
| AK216505.1 | - | 5 | 284 | |||
| BC005610.1 | - | 10 | 752 | |||
| BC025448.1 | - | 8 | 517 | |||
| BC038821.1 | - | 13 | 1028 | 3990023 | ||

Tada2a
Genomic Map