siRNA Details
| Gene Symbol: Tada2a | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_172562.3 | NP_766150.1 | 10, 11 | 819 | ||
| GenBank mRNA | AK157938.1 | - | 10, 11 | 812 | ||
| AK216505.1 | - | 2, 3 | 78 | |||
| BC005610.1 | - | 7, 8 | 546 | |||
Please enter the information below and press OK to send your cart to Core Services for purchase.

Tada2a| Gene Symbol: Tada2a | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_172562.3 | NP_766150.1 | 10, 11 | 819 | ||
| GenBank mRNA | AK157938.1 | - | 10, 11 | 812 | ||
| AK216505.1 | - | 2, 3 | 78 | |||
| BC005610.1 | - | 7, 8 | 546 | |||