siRNA Details
| Gene Symbol: Tada2a | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_172562.3 | NP_766150.1 | 4 | 265 | ||
| GenBank mRNA | AK142262.1 | - | 4 | 297 | ||
| AK157938.1 | - | 4 | 258 | |||
| BC038821.1 | - | 4 | 268 | 3990023 | ||
Please enter the information below and press OK to send your cart to Core Services for purchase.

Tada2a| Gene Symbol: Tada2a | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_172562.3 | NP_766150.1 | 4 | 265 | ||
| GenBank mRNA | AK142262.1 | - | 4 | 297 | ||
| AK157938.1 | - | 4 | 258 | |||
| BC038821.1 | - | 4 | 268 | 3990023 | ||